Imprimer
Chemical resistant overalls, Tyvek® 400 DualCombi
Fournisseur: DuPont
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
TD0125MWH00 TD0125SWH00 TD0125LWH00 TD0125XLWH00 TD01252XWH00 TD01253XWH00 TD01254XWH00
DUPOTD0125MWH00EA 220 EUR
DUPOTD0125MWH00 DUPOTD0125SWH00 DUPOTD0125LWH00 DUPOTD0125XLWH00 DUPOTD01252XWH00 DUPOTD01253XWH00 DUPOTD01254XWH00
Chemical resistant overalls, Tyvek® 400 DualCombi
Vêtements à usage unique
These collared overalls feature Tyvek® at the front and a large breathable spunbound polypropylene back panel. Ideal for use in environments where comfort is important and exposure risk is limited to the front.
  • Elasticated wrists and ankles
  • Thumb hole on sleeve
  • Elasticated waist (stitched-in)
  • Nylon zipper with flap
  • With stitched external seams for enhanced protection against penetration from the outside to the inside of the garment
Ideal for use in environments where comfort is important and exposure risk is limited to the front. Developed with end users to answer their needs for superior frontal protection while sustaining high ventilation and breathability in the back.
Tyvek® 400 DualCombi is designed for streneous jobs done in warm environment that demand comfort while helping to provide protection from frontal exposure. Common applications are paint spraying and jobs involving composite materials and utilities.
Certifications: Type PB[6]; Cat. III
Cdt: 25 per box, bulk packed.