Imprimer
Human bone marrow-mesenchymal stem cells (hBM-MSC), CELLvo™
Marques : STEMBIOSYS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BM-100-000 BM-100-XF-6WP BM-100-XF-T75 BM-100-XF-T150
STBSBM-100-000EA 826 EUR
STBSBM-100-000 STBSBM-100-XF-6WP STBSBM-100-XF-T75 STBSBM-100-XF-T150
Human bone marrow-mesenchymal stem cells (hBM-MSC), CELLvo™
Cellules
CELLvo™ Human Bone-Marrow Mesenchymal Stem Cells are adult multipotent stem cells cryopreserved at passage 2.
  • Highly proliferative
  • Smaller cell size
  • Higher percentage of SSEA4+ cells
  • >20 population doublings
CELLvo™ Bone Marrow - Mesenchymal Stem Cells (hBM-MSC) have been isolated and cultured under xeno free conditions on CELLvo™ XF-Matrix. CELLvo™ hBM-MSCs proliferate more quickly, have a smaller cell size and show a higher percentage of SSEA4 (+) cells as compared to BM-MSCs isolated and cultured on other substrates.
Tested Applications: Differentiation potential, flow cytometry for phenotype, bone formation.
Informations de commande: CELLvo™ Human Bone Marrow - Mesenchymal Stem Cells (hBM-MSC) are derived from a single donor and cryopreserved at P2. CELLvo™ hBM-MSCs can be ordered as a single vial of cells or as a kit to include CELLvo™ XF Matrix.
Informations de livraison: A vial of cells will include >1 M CELLvo™ hBM-MSCs. A kit will contain a single vial of hBM-MSCs cryopreserved at passage 2 and a sleeve of CELLvo™ XF Matrix (6 well plates, T 75 flasks or T 150 flasks). Additional CELLvo™ XF Matrix can be purchased separately.
Cdt: CELLvo™ hBM-MSCs arrive on dry ice and may include a sleeve of CELLvo™ XF Matrix. hBM-MSCs should be stored in liquid nitrogen upon arrival. CELLvo™ XF Matrix should be stored at 2 to 8 °C upon arrival. Avoid extended exposure to light.
Attention: CELLvo™ products are not for human use. These products should not be used for diagnostic or therapeutic purposes. Handle in accordance with established bio-safety practices.