Imprimer
Sustainable 2-in-1 sterile flat mop covers, Revolve™
Fournisseur: Texwipe
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
STX1720 STX1721
129-1775EA 1490 EUR
129-1775 129-1776
Sustainable 2-in-1 sterile flat mop covers, Revolve™
Serpillières
Revolve™ 2-in-1 mop covers, the first sustainable covers made for the cleanroom industry, are made from 100% upcycled polyester. Revolve™ mop covers contribute to sustainability goals, reducing the amount of post-consumer water bottles going into landfills and reducing the level of carbon emissions.
  • Mop and clean large and small surfaces
  • Apply and remove disinfectants
  • Spill control
  • 100% upcycled polyester
  • Clean with solvents such as isopropyl alcohol (IPA), ethanol, acetone and degreasers
  • Appropriate for use with temperatures less than 205 °C (400 °F)
The integrated polyester cover/pad design allows the replacing of covers and pads in a one-step process. The integrated design offers high absorption capacity, which is ideal for spill control, cleaning, and solution application. The covers easily fit over the mop head, permitting a quick change. The integrated polyester pad ensures the mop cover conforms to surfaces for consistent contact and cleaning. Revolve™ processing provides low levels of ions, NVRs (non-volatile residues), particles and fibres for use in critical cleaning applications and environments. Autoclave safe. Individually lot coded for traceability and quality control.
Certifications: Sterile wipes are compatible for use in GMP grade A.