Imprimer
Grounding and adapters, SymLine®
Fournisseur: S.C.A.T.
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
106415 106410 107632 106423 106409 106408 108079 106417 106434 121211 108176 106510 106539 106584 108272 108262 106523 108270
554-4360EA 46.5 EUR
554-4360 554-1768 554-1848 554-4364 554-4359 554-4358 SCTV108079 SCTV106417 SCTV106434 SCTV121211 SCTV108176 SCTV106510 SCTV106539 SCTV106584 SCTV108272 SCTV108262 SCTV106523 SCTV108270
Grounding and adapters, SymLine®
Raccords et connecteurs
Dovetail the SymLine® system to suit specific requirements. This product selection includes several useful ancillaries.
  • SymLine® offers a suitable adapter for every common thread, to eliminate problems connecting different thread types
  • Many useful accessories allow users to configure the SymLine® system according to specific needs
  • All accessories are eclectically dissipative and approved for use in laboratories with explosion hazards
Dispose of liquid HPLC waste safely. SymLine® is quickly installed without drilling holes or making technical changesEn savoir plus
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable