Imprimer
Cleanroom gloves, nitrile, non sterile, white, smooth finish, PUREZERO® HG3
Fournisseur: Halyard
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
CLN3231LG CLN3231XL CLN3231MD CLN3231SM CLN3231XS
112-6519EA 560 EUR
112-6519 112-6522 112-6520 112-6521 112-6523
Cleanroom gloves, nitrile, non sterile, white, smooth finish, PUREZERO® HG3
Gants
High quality gloves are designed to exact standards to meet the requirements of a contamination controlled environment.
  • High quality nitrile
  • Non sterile
  • Accelerator-free formula reduces risk of type IV allergies
  • For use in cleanroom environments (ISO 5/6/7/8, grade B/C/D aseptic)
  • REACH Annex XVII
Offering the right glove for the right controlled environment, Halyard PUREZERO® provides a cleanroom glove with guaranteed low particulate counts and endotoxin levels due to repeated washes in deionised water and chlorination. Nickel-, powder-, silicon- and latex-free, with no phthalates. These gloves are formulated with zero presence of these hazardous traditional accelerators: Sulphur, thiurams, thiazoles, guanidine and carbamates (ZDEC or ZDBC). All backed up by a vertically integrated supply chain.
Certifications: These cleanroom gloves are certified as PPE category III per the Regulation (EU) 2016/425. The applicable standards are:
• EN ISO 374-1:2016 Type C K - dangerous chemicals and microorganisms (permeation time)
• EN ISO 374-2:2019 - dangerous chemicals and microorganisms (determining resistance to penetration holes/defects)
• EN ISO 374-4:2019 - resistance to degradation by chemicals
• EN ISO 374-5:2016 - microorganism and virus protection
• EN 420:2003 + A1:2009 - general requirements for protective gloves
• EN 16523-1:2015 + A1:2018 - permeation of chemicals (+IPA 70%**)
• ISO 16604:2004 - contact with blood and body fluids
• ASTM D6978 - permeation testing on chemotherapy drugs (14 drugs)
Cdt: Gloves are double-bagged, per 100 pieces.