Imprimer
Biemer vessel spring scissors
Fournisseur: FINE SCIENCE TOOLS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
15032-13 15032-08
FSCI15032-13EA 670 EUR
FSCI15032-13 FSCI15032-08
Biemer vessel spring scissors
Ciseaux
These stainless steel scissors are designed to securely cut vessels without slipping.