Imprimer
Vannas spring scissors, 2,5 mm cutting edge
Fournisseur: FINE SCIENCE TOOLS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
15000-08 15001-08 15002-08
FSCI15000-08EA 384 EUR
FSCI15000-08 FSCI15001-08 FSCI15002-08
Vannas spring scissors, 2,5 mm cutting edge
Ciseaux
These stainless steel scissors have been designed to maximise visibility under the microscope by eliminating unnecessary bulk in the blades.
A short taper makes these scissors ideal for small, precise cuts.