Imprimer
Syringe filters, prefilter, sterile, Whatman™ Anotop Plus
Fournisseur: Whatman products (Cytiva)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6809-3112 6809-3102 6809-4102
516-1136EA 725 EUR
516-1136 516-1135 514-8036
Syringe filters, prefilter, sterile, Whatman™ Anotop Plus
Filtres Filtres-seringue
Cytiva's Whatman Anotop Plus sterile syringe filter with glass microfiber prefilter is compatible with most organic and aqueous solutions for use in a wide variety of applications.
  • Contains an inorganic Anopore membrane and is available in three pore sizes
  • Polypropylene syringe filters comes in two diameters depending on sample volume
  • Integrated glass microfiber prefilter aids in filtration of difficult samples
  • Recyclable after use
Anotop Plus filters are manufactured with an Anopore membrane suitable for many applications, including capillary electrophoresis, UV/Vis sample preparation, protein separation, and HPLC sample preparation.