Imprimer
Ready-to-use tyramide amplification buffer, 1X
Marques : Biotium
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
22027-100ML 22027-25ML
22027-100MLEA 696 EUR
22027-100ML 22027-25ML
Ready-to-use tyramide amplification buffer, 1X
Tampons Tampons biologiques
For simple, optimised tyramide amplification of immunostaining in cells or tissue sections. No need to add hydrogen peroxide before use.
  • Ready-to-use
  • Simply dilute tyramide probe in buffer and proceed to signal development
  • Compatible with tyramides from Biotium or other suppliers
The Tyramide Amplification Buffer Plus (BTIU22029), a improved buffer for tyramide signal amplification provides significantly improved brightness, specificity, and sensitivity over this original buffer.
Simply mix the tyramide compound with the buffer to make a working solution for signal amplification. Although specifically optimised for our CF® Dye-, Biotin XX-, and DNP-tyramide products, Ready-to-Use Tyramide Amplification Buffer is also compatible with tyramide probes from other manufacturers.