Imprimer
CF® Dye Alkyne
Marques : Biotium
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
92086 92087 92088 92089 92090 92091 92093 92095 96004 96005 96006
92086.EA 485 EUR
92086. 92087. 92088. 92089. 92090. 92091. 92093. 92095. 96004. 96005. 96006.
CF® Dye Alkyne
Systèmes de liaison et de marquage des protéines
CF® Dye alkynes react with azide groups via copper-catalysed bioorthogonal cycloaddition. Can be used to fluorescently detect or label azide groups on target molecules. CF® Dye alkynes can also be used as building blocks to form fluorescent polymers.
  • Fluorescent detection or labelling azide groups on target molecules
  • Can be used as fluorescent monomers to form dye-labelled polymers
  • Bright, photostable and water-soluble CF® Dyes are excellent options for fluorescent labelling
CF® Dye alkynes can react with azide groups to form 1,2,3-triazole via 1,3-dipolar Huisgen copper-catalysed cycloaddition. They can also be used as monomeric building blocks to copolymerise with other monomers to form fluorescent polymers.
CF® Dyes are of next-generation fluorescent dyes that have improved brightness, photostability and water solubility compared to Alexa Fluor®, DyLight®, and other fluorescent dyes.