Imprimer

Electric autoclaves for sterilisation, vertical table top, Presoclave II

Fournisseur: SELECTA
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

4002136 4002135
481-0600EA 0 EUR
481-0600 481-0599
Electric autoclaves for sterilisation, vertical table top, Presoclave II
Stérilisateurs Autoclaves
Electrical autoclaves for sterilisation. External case made from AISI 304 stainless steel. Sterilisation chamber and door made from AISI 316 stainless steel.

  • INCOLOY stainless steel heater elements, hard waring, resistant to corrosion and high temperatures
  • Automatic purge to atmosphere
  • Lid, six point closure
  • Control panel with: illuminated alarm, door open and end of cycle indicator, pressure gauge, temperature and time display

Electronic control of time and temperature with digital display temperature range from 115 to 134 °C (0,62 to 2 bar). The safety over pressure valve prevents over pressure above the maximum pressure limit. Safety thermostat and water level disconnect power to the heater if insufficient water or over pressure. The safety lid prevents the lid being opened if the chamber is pressure.

Informations de commande: Does not include baskets, order separately
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR