Imprimer
Mammalian TAP system, interPlay
Marques : AGILENT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
240102 240106
AGLS240102EA 789 EUR
AGLS240102 AGLS240106
Mammalian TAP system, interPlay
Vecteurs, plasmides et librairies
The InterPlay Mammalian TAP system allows you to recover interacting proteins from mammalian cells.
  • Protease digestion unnecessary to recover intact interacting protein partners since SBP and CBP elute easily from their affinity resins
  • Exceptionally pure protein for downstream analysis
This method is based on expression of a protein of interest fused to two affinity tags: a streptavidin binding peptide (SBP) and a calmodulin binding peptide (CBP). Tandem affinity purification yields your tagged protein and interacting proteins using gentle washing and small molecule elution conditions.
Attention: For research use only. Not for use in diagnostic procedures.