Imprimer
Paq5000 DNA polymerase
Marques : AGILENT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
600684 600862 600864
AGLS600684EA 360 EUR
AGLS600684 AGLS600862 AGLS600864
Paq5000 DNA polymerase
Réactifs pour acides nucléiques End-point PCR Enzymes and Kits
Paq5000 is capable of quickly and robustly amplifying up to 6 kB genomic DNA targets.
  • Equal to better yield with fast cycling conditions
  • Robust, cost-effective
  • Pre-optimised reaction buffers included
  • Low cost hot start formulation
With a shortened extension rate of 30 sec/kB and robust yields, Paq5000 is a Taq alternative that can be substituted easily into many Taq-based procedures.
Paq5000 includes the ArchaeMaxx factor, improving the yield of products by overcoming dUTP poisoning, which is caused by dUTP accumulation during PCR through dCTP deamination. Once incorporated, dU-containing DNA inhibits the enzyme and the ArchaeMaxx factor functions as a dUTPase, converting poisonous dUTP to harmless dUMP and inorganic pyrophosphate.
Attention: For research use only. Not for use in diagnostic procedures.