Imprimer
VWR®, Lymphocyte separation media
Marques : VWR Chemicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
392-0431EA 24.9 EUR
392-0431 392-0432
VWR®, Lymphocyte separation media
Milieux de culture cellulaire
Lymphocyte separation media is a density gradient medium for the isolation of mononuclear cells from peripheral blood, cord blood and bone marrow.
  • Like Ficoll™, VWR® lymphocyte separation media has:
  • An optimised density of 1,077 g/ml
  • Granulocytes and erythrocytes will sediment under the media layer
  • Mononuclear cells will be isolated to top of the layer
Designed for a simple, rapid isolation by centrifugation of lymphocytes from whole blood, once diluted and treated with anti-coagulant or defibrinating agent.