Imprimer
Seals for GL screw caps, DURAN®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
292480805 292481107 292481304 292481904 292482806
215-2299EA 9.5 EUR
215-2299 215-2300 215-2308 215-2309 215-2310
Seals for GL screw caps, DURAN®
Capuchons Bouchons pour flacons
Silicone with PTFE coating.
  • Thermal resistance: 130 °C (steam) and 200 °C (heat)
  • Good chemical resistance thanks to the PTFE coating
Suitable for GL screw caps, premium and tamper-evident caps.