Imprimer
Adapter with ground joint and swivelling fittings with GL- thread
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
D692-64 D692-34 D692-44 D692-24 D692-54
201-0270EA 167 EUR
201-0270 201-0268 201-0269 201-0275 BOHLD692-54
Adapter with ground joint and swivelling fittings with GL- thread
Adapters Adapters, Conical Ground Joints
Adapter with screw fitting made from PPS, cone joint body made from PTFE.
  • Very good chemical resistance
  • Temperature resistant from –50 to +250 °C
  • Suitable for vacuum applications
For the insertion of probes, tubes and hoses into vessels with standard ground joints. To prevent contact with agitator shafts and so on. Sensors, tubes or hoses are initially guided through the threaded connection. The adapter is then inserted in the joint and the inserted element (tube or probe) is secured in the required position.