Imprimer
Glass beakers, low form, Super Duty, DURAN®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
211074104 211074807 211075409 211076302 211077307 211072909 211073605
214-0205EA 24 EUR
214-0205 214-0206 214-0207 214-0208 214-0209 214-0199 214-0204
Glass beakers, low form, Super Duty, DURAN®
Béchers
DURAN®, borosilicate glass 3.3, clear, with spout.
  • Higher mechanical stability compared with standard glass beakers due to reinforced sturdy rim
  • Even distribution of wall thickness
  • Very good chemical resistance, good thermal resistance
With easily legible graduation and labelling area. It is recommended that you heat Super Duty glass beakers slowly and evenly to prevent thermal stresses in the glass. For work at very high temperatures, or with fast changes in temperature, standard DURAN® beakers should be used, as these offer excellent resistance to temperature changes. However, mechanical stability is limited with these products in comparison with the Super Duty beakers.
With printed Retrace Code (batch identification).