Imprimer
Terrestrial (Cypripedium) orchid medium, contains 600 mg/L calcium nitrate
Marques : Apollo Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PMT842A-10X1L PMT842B-10L PMT842C-50L
APOSPMT842A-10X1LEA 32.2 EUR
APOSPMT842A-10X1L APOSPMT842B-10L APOSPMT842C-50L
Terrestrial (Cypripedium) orchid medium, contains 600 mg/L calcium nitrate
Milieux de culture cellulaire
Contains 600 mg/L calcium nitrate and 200 mg/L casein; without ammonium nitrate.
  • Plant tissue culture tested