Imprimer
NGS FFPE QC kit
Marques : AGILENT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
G9700A G9700B
AGLSG9700AEA 302 EUR
AGLSG9700A AGLSG9700B
NGS FFPE QC kit
Tests Tests à l'acide nucléique
The Agilent NGS FFPE QC kit is a qPCR-based assay that enables functional DNA quality assessment of input DNA prior to preparation of next generation sequencing (NGS) libraries.
  • A powerful qPCR-based assay that enables functional DNA quality assessment of input DNA prior to preparation of NGS libraries
  • Efficiently assess both DNA integrity and accurate quantitation of amplifiable templates prior to library preparation
  • Accurately qualify and quantify amplifiable DNA in challenging formalin-fixed, paraffin-embedded tissue samples
  • Includes an optimised low input library prep workflow for improved complexity and target coverage
  • Provides complete cancer research solutions from sample to data
  • Includes a full qPCR-based QC and Quant kit with reagents for processing samples, including reference and standard DNAs in triplicate
The Agilent NGS FFPE QC kit is a qPCR-based assay that enables functional DNA quality assessment of input DNA prior to preparation of next generation sequencing (NGS) libraries. The formalin-fixed, paraffin-embedded tissue QC kit enables assessment of the integrity of DNA. It also ensures accurate quantitation of amplifiable template going into library preparation.
Formalin-fixed, paraffin-embedded tissue represents a valuable sample source for molecular cancer research but is highly challenging to analyse. Our FFPE DNA NGS QC helps achieve this, enhancing research and analysis efficiency. In addition, optimisations to the SureSelectxt workflows based on the sample quality score are enabled.