Imprimer
VWR®, Test Tube Racks, Mega, for 18 to 20 mm Tubes
Fournisseur: VWR
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
VWR®
525-1595EA 78.7 EUR
525-1595 525-1594
VWR®, Test Tube Racks, Mega, for 18 to 20 mm Tubes
Portoirs Tube Racks
These fully-assembled, polypropylene Mega Racks are great for high throughput labs requiring large sample storage.
  • Stores a high quantity of samples eliminating the need for multiple racks
  • Alphanumeric grid for quick location and retrieval of tubes
Made of sturdy, lightweight material for easy transport.