Imprimer
Transfection reagents, GeneJammer
Marques : AGILENT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
204130 204132
AGLS204130EA 562 EUR
AGLS204130 AGLS204132
Transfection reagents, GeneJammer
Transfection Reagents
The GeneJammer transfection reagent is a proprietary formulation of novel polyamines ideal for transient or stable transfections in a wide variety of cell types.
  • Fast and easy to use, no media changes required
  • Enables high level adenovirus transduction in the absence of an adenovirus specific receptor
  • Works with media containing antibiotics, growth factors and/or other supplements
This transfection reagent is effective in both serum-containing medium and serum-free medium, offering high efficiencies with minimal cytotoxicity.
1.33 mg/ml reagent in 80% ethanol.
Attention: For research use only. Not for use in diagnostic procedures.