Imprimer
Flat head mop wringer, MegaWringer™
Fournisseur: MICRONOVA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
W-9 W-92 W-9B W-94
129-0064EA 465 EUR
129-0064 141-0107 216-1491 216-2571
Flat head mop wringer, MegaWringer™
Serpillières
A stainless steel flat head mop wringer with a unique design that leverages the mop handle to exert downward wringing pressure, doing away with the need to bend and press while cleaning.
  • Down-press wringer, ideal for consistent wringing
  • One hand action, easy to use with minimal pressure
  • Guards against operator wrist and back strain
  • Unit is compact, easily transported and stored, and weighs less than 5 pounds
  • Electropolished stainless steel, with moulded polyketone components
  • Pair with Slim T™ buckets for cart-free use
  • Holds up to strong disinfectants
  • Autoclavable
The Micronova MegaWringer™ flat head mop wringer features a simple, sturdy design that leave mops damp enough to effectively disinfect surfaces, while wringing out excess liquid. Designed to be used with Slim T™ buckets in BucketBinder™ systems or Slim T™ cart systems.