Imprimer
Terrestrial (Cypripedium) orchid medium
Marques : Apollo Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PMT839A-10X1L PMT839B-10L PMT839C-50L
APOSPMT839A-10X1LEA 46.8 EUR
APOSPMT839A-10X1L APOSPMT839B-10L APOSPMT839C-50L
Terrestrial (Cypripedium) orchid medium
Milieux de culture cellulaire
Contains 400 mg/L calcium nitrate and 400 mg/L Casein; WITHOUT ammonium nitrate.