Imprimer
NMR tubes with Bruker® SampleJet® caps, SP Wilmad-LabGlass
Fournisseur: SP Industries
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
WG-1000-4-SJ WG-3000-4-SJ WG-3000-7-SJ WG-1000-7-SJ
634-1185EA 248 EUR
634-1185 634-1187 634-1188 634-1186
NMR tubes with Bruker® SampleJet® caps, SP Wilmad-LabGlass
Tubes Tubes RMN
These Wilmad-LabGlass NMR tubes are supplied with authentic closed Bruker® SampleJet® caps without code.
  • Camber and concentricity values listed are the Total Indicator Reading (TIR)
  • One of the best OD tolerances in the industry
  • Guaranteed to fit tightly in spinner turbine
  • Made from ASTM E438 Type 1 Class B glass
  • 100% inspected for surface defects and physical dimension to ensure the success of your experiments
Designed for routine use in most low to mid field NMR spectrometers.