Imprimer
Syringe filters, SupaTop
Fournisseur: Gilson
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
GILSANP2545EA 97 EUR
GILSANP2545 GILSANP2522 GILSANM2522C GILSANR1322 GILSANR1345 GILSANV2522C GILSANR1345D GILSANM1322 GILSANM1345 GILSANP2522C GILSANV2522 GILSANR2545C GILSANM1345D GILSANV1345D GILSANN2522C GILSANV2545 GILSANN2522 GILSANV2545C GILSANN1322D GILSANR2545 GILSANP1345D GILSANN2545 GILSANM2545C GILSANP1322 GILSANP1345 GILSANR2522 GILSANR1322D GILSANP2545C GILSANM2522 GILSANM2545 GILSANM1322D GILSANV1345 GILSANV1322 GILSANP1322D GILSANV1322D GILSANR2522C GILSANN2545C GILSANN1322 GILSANN1345 GILSANN1345D
Syringe filters, SupaTop
Filtres Filtres-seringue
SupaTop syringe filters offer reliable and cost-effective solutions for particles removal and clarification needs.
  • Body made of pigment-free PP
  • Certified for low levels of UV-absorbing extractables
A large variety of membranes are available for all applications: MCE, Nylon, PTFE, PES or PVDF.