Imprimer
Hermetic Tamper Evident Buckets, Round
Fournisseur: LAMAPLAST
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1113 1139 1138 1135 1133 1132 1127 1117 1119
LAMA1113EA 7.1 EUR
LAMA1113 LAMA1139 LAMA1138 LAMA1135 LAMA1133 LAMA1132 LAMA1127 LAMA1117 LAMA1119
Hermetic Tamper Evident Buckets, Round
Godets
Robust PP bucket with ergonomic handle. It features pressure lid with opening-off seal.
  • Flat bottom ensures total emptying and easy cleaning
  • Especially suitable in cosmetic, pharma, paint or dye industries
  • Food contact material approved
Hot filling possible up to 95 °C.
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable