Imprimer
VWR®, Accessories for VWR® Professional and Advanced Hotplate Stirrers
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
442-1297EA 348 EUR
442-1297 442-1298 442-1279 442-1280 442-1281 442-1282 442-1283 442-1284 442-1285 442-1286 442-1287 442-1288 442-1289 442-1290 442-1291 442-1292 442-1293 442-1294 442-1295 442-1296
VWR®, Accessories for VWR® Professional and Advanced Hotplate Stirrers
Agitateurs Agitateurs magnétiques chauffants
The VWR® Multi-Sample Reaction Station enables heating and stirring of one size or up to five multiple sample sizes, all on one unit. The base plate has a centrally located threaded opening to accommodate a 13 mm diameter support rod.
  • Heat and stir a variety of samples simultaneously on one hotplate
  • Uni and sectional blocks accommodate 9 different sample sizes
  • Anodised aluminum with close block to tube contact ensures optimal transfer of heat
Blocks are made of anodised aluminum to provide superior temperature stability and heat transfer. Each block has a thermometer well for measuring temperature. The base plate is designed to fit VWR® round top hotplate stirrers or other 135 mm diameter top models. Safety handles are available to aid in the safe removal of the blocks or base plate.