Imprimer
Perfect Match PCR enhancers
Marques : AGILENT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
600129 600130
AGLS600129EA 482 EUR
AGLS600129 AGLS600130
Perfect Match PCR enhancers
Réactifs pour acides nucléiques qPCR/RT-qPCR Enzymes and Kits
Increases specificity and yield of primary PCR amplification products.
  • Minimises formation of poorly matched primer-template complexes
  • Add to genomic PCR reactions to destabilise many mismatched primer-template complexes
Explore how Perfect Match PCR enhancer increases the specificity and yield of primary PCR amplification products. When added to genomic in vitro amplification reactions, it destabilises numerous mismatched primer-template complexes that would otherwise result in a heterogeneous population of amplified molecules.
Attention: For research use only. Not for use in diagnostic procedures.