Imprimer
J.T.Baker®, Robotic Tips Type OLA
Fournisseur: Avantor
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
49044-0000EA 127 EUR
49044-0000 49044-0100
J.T.Baker®, Robotic Tips Type OLA
Pipette Tips
Suitable for use on OLA 2500 and AutoMate 1250/2550 instruments. Available in conductive version, with or without filter.
  • 140 tips are sealed in one optimized rack
  • Working volumes of 20 to 1200 µl
The validation of the tips on the instrument for specific applications is within the responsibility of the user.