Imprimer
PCR cloning kits, StrataClone
Marques : AGILENT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
240205 240207 240218
AGLS240205EA 458 EUR
AGLS240205 AGLS240207 AGLS240218
PCR cloning kits, StrataClone
Réactifs pour acides nucléiques Cloning Kits and Ligation
The StrataClone PCR cloning kit allows high-efficiency, 5-minute cloning of PCR products at room temperature, using the efficient DNA rejoining activity of DNA topoisomerase I and the DNA recombination activity of Cre recombinase.
  • High efficiency yields >95% insert positive clones
  • Clone both long and short amplicons (500 bp to 9,2 Kb)
  • With kanamycin and ampicillin resistance markers
These three kits are available for both blunt-end and UA cloning. Additionally, combined the highest accuracy in PCR amplification with easy, reliable, and affordably priced topoisomerase-based cloning in StrataClone Ultra PCR cloning system, which includes the Pfu Ultra II Fusion Hotstart DNA Polymerase.
StrataClone PCR cloning kit: High efficiency UA PCR cloning featuring DNA topoisomerase I and Cre recombinase technologies.
StrataClone blunt PCR cloning kit: High efficiency blunt-end PCR cloning, features DNA topoisomerase I and Cre recombinase technologies.
StrataClone ultra blunt PCR cloning kit: High efficiency blunt-end PCR cloning, features DNA topoisomerase I and Cre recombinase technologies, Kit includes PfuUltra II for high fidelity PCR amplification.
Attention: For research use only. Not for use in diagnostic procedures.