Imprimer
Accessories for FlexTube and FlexTube Pro, SymLine®
Fournisseur: S.C.A.T.
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
106558 106562 106579 106591
SCTV106558EA 20.2 EUR
SCTV106558 SCTV106562 SCTV106579 SCTV106591
Accessories for FlexTube and FlexTube Pro, SymLine®
Tuyaux Tuyaux en polyéthylène
These flexible tubes made from PTFE can be used in safety cabinets observing the specified maximum dimension for drillings. They are the ideal solution for installation in confined spaces since they can be placed easily into clearances and provide the flexibility required in safety cabinets with pull-out containers.
  • Made of PTFE for best possible chemical resistance
  • Adjustable and easy to install; shorten tube as required, then mount the optimized screw connection and sealing
  • Electrostatic conductive <106 (max) Ω
  • 16,9 or 19,8 mm OD
  • Available in 4 different standard lengths: 600, 1200, 1500, and 2500 mm
FlexTube with inner wall spiral or or FlexTubePro Inner wall smooth. The two most flexible and adaptable tubing systems for the safe disposal of liquid HPLC waste. Flexible, expandable, chemically resistant PTFE tubing with a large bending radius. SymLine® FlexTube Pro with unruffled inner wall prevents waste and residues in tubes.