Imprimer
Serum-free media, HyClone™ SFMTranfx-293 media
Marques : HyClone products (Cytiva)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SH30860.03 SH30860.04 SH30861.01
HYCLSH30860.03EA 361 EUR
HYCLSH30860.03 HYCLSH30860.04 HYCLSH30861.01
Serum-free media, HyClone™ SFMTranfx-293 media
Milieux de culture cellulaire
HyClone™ SFMTransfx-293 is a serum-free, animal derived component-free medium designed to support the growth of HEK 293 cultures and promote transfection using lipofection or similar methods. Manufactured according to cGMP guidelines.
  • Supports high transfection efficiency
  • Metabolically designed for high cell yield and recombinant protein/vector production
  • Allows for direct or sequential adaptation
HyClone SFMTransfx-293 Media is a regulatory-friendly medium developed through the Metabolic Pathway Design to support high transfection efficiency, productivity and cell density in suspension cultures.
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable