Imprimer
Polystyrene centrifuge tubes, PerformR®
Fournisseur: LABCON
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
3115-870-028-9 3130-345-008-9
3115-870-028-9EA 284 EUR
3115-870-028-9 3130-345-008-9
Polystyrene centrifuge tubes, PerformR®
Tubes Tubes à centrifuger
Polysterene tubes are made of crystal clear resin and have bold etched graduations. They can be used for some culture applications that require a glass clear material.
  • ATP-, RNase/Dnase-, and endotoxin-free (non-pyrogenic)
  • Attached flat screw caps with a polyethelene liner
  • Molded graduations and writing surface
  • Polystyrene USP Class VI resin, free of DiHEMDA, oleamide, and BPA
  • Not freezable, not autoclavable
Caps feature a three-ply, co-extruded liner consisting of a foamed white low density polyethelene (LDPE) core sandwiched between two solid layers of LDPE, which contains high density polyethelene (HDPE) for improved stiffness.