Imprimer
Revolve™ Sustainable Pre-Wetted Cleanroom Wipes
Fournisseur: Texwipe
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
TX1704P TX1709P TX1712P
115-2717EA 473 EUR
115-2717 115-2719 115-2731
Revolve™ Sustainable Pre-Wetted Cleanroom Wipes
Lingettes Lingettes pour salles propres Lingettes pour salles propres, présaturées
Revolve™ sustainable wipes are made for the cleanroom industry and are composed of 100% upcycled polyester material with a sealed edge, processed on Texwipe’s fully automated manufacturing system. Revolve™ wipes contribute to sustainability goals, reducing the amount of post-consumer water bottles going into landfills and reducing the level of carbon emissions.
  • Wipe and clean surfaces, equipment and parts
  • Apply and remove lubricants, adhesives, residues and other solutions including disinfectants
  • Clean with solvents such as isopropyl alcohol (IPA), ethanol, acetone and degreasers
Revolve™ processing provides low levels of ions, NVRs (non volatile residues), particles, and fibres for use in critical cleaning applications and environments. They are designed for high sorption capacity, which is ideal for spill control, cleaning, and solution application.
Revolve™ pre-wetted products are pre-wet with 0,2 µm filtered USP-grade 70% IPA/30% DIW. Pre-wetted wipes are packed in easy to use, recloseable slider bags, reducing solution evaporation that preserves the consistent wipes wetness level.
Post-consumer bottles made from virgin polyester are reused and upcycled through the REPREVE® recycling process to make the yarn used in the Revolve™ products, keeping these bottles from undergoing disposal in landfills.
Certifications: Meet USP and USP wipe requirements. Non-sterile wipes are compatible for use in ISO class 3 to 7 / GMP grade B to D.
Attention: *The product(s) on this page are outside the scope of the EU Directive 2019/904.