Imprimer
DECON-Clean® cleaning agents
Fournisseur: VELTEK ASSOCIATES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
DC-01-E DC-03-4Z-E DC-02-E
VELTDC-01-EEA 511 EUR
VELTDC-01-E VELTDC-03-4Z-E VELTDC-02-E
DECON-Clean® cleaning agents
Produits nettoyants Détergents et Nettoyants de Surfaces
Ideal for washable, nonporous surfaces such as walls, ceilings, floors, and stainless steel items, this cleaning agent is designed to remove residues left behind by disinfecting agents
DECON-Clean® is filtered at 0.2µm, gamma irradiated, and double-bagged. SimpleMix® one-step residue remover dilutes to 1:128 in Water for Injection. Sterile options are USP tested and delivered with a lot-specific certificate of irradiation and certificate of analysis. Traceable sterile cleaning agent comes with an LAL testing analysis and sterility report.