Imprimer
Phosphorus CRM, standard solutions, Supelco®
Marques : Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1.25046.0100 1.25047.0100 1.25048.0100 1.25049.0100
1.25046.0100EA 59.7 EUR
1.25046.0100 1.25047.0100 1.25048.0100 1.25049.0100
Phosphorus CRM, standard solutions, Supelco®
Étalons Étalons anioniques Standards de phosphate
  • Diluted standard solutions that are ready-to-use for accurate measurements
  • Traceable to SRM from NIST and USP
  • Full certificates of analysis with comprehensive quality documentation
  • Information on concentration and expanded measurement uncertainty
  • Save time and minimize errors
Method used for Uncertainty Measurement (Accuracy in the table below)= Photometry
Certifications: Traceable to SRM from NIST and USP.