Imprimer
Coronavirus and SARS-CoV-2 RT real-time PCR kits, GSD NovaPrime®
Marques : GOLD STANDARD DIAGNOSTICS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PCOV6034 PCOV6036 PCOV6037
GSDCPCOV6034EA 2810 EUR
GSDCPCOV6034 GSDCPCOV6036 GSDCPCOV6037
Coronavirus and SARS-CoV-2 RT real-time PCR kits, GSD NovaPrime®
Systèmes de diagnostic clinique Réactifs et Kits de diagnostic clinique
This RT-PCR assay is a highly sensitive Multiplex Real-Time PCR with CE-IVD marking for the direct qualitative pathogen detection of SARS-CoV-2 genomic RNA in respiratory samples.
  • Approximately 1 hour turnaround time
  • Specimen types: nasal wash/swab, nasopharyngeal wash/swab, oropharyngeal swab, BAL, pharynx gargle solution, and saliva
  • Excellent performance with a 100% NPA, PPA
  • Extraction/inhibition control included
The assay simultaneously detects two target sequences in the N gene.
Certifications: CE-IVD marked.