Imprimer
TOC Standard Solutions, Certified Reference Materials, Ready-to-Use, for Photometry, Supelco®
Marques : Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1.32246.0100 1.32247.0100 1.32248.0100 1.32249.0100 1.32251.0100 1.32252.0100 1.32253.0100
1.32246.0100EA 72.4 EUR
1.32246.0100 1.32247.0100 1.32248.0100 1.32249.0100 1.32251.0100 1.32252.0100 1.32253.0100
TOC Standard Solutions, Certified Reference Materials, Ready-to-Use, for Photometry, Supelco®
Étalons Étalons pour l'analyse environnementale
  • Diluted standard solutions that are ready-to-use for accurate measurements
  • Traceable to SRM from NIST and USP
  • Full certificates of analysis with comprehensive quality documentation
  • Information on concentration and expanded measurement uncertainty
  • Save time and minimize errors
Method used for Uncertainty Measurement (Accuracy in the table below)= TOC-Analyzer
Certifications: Traceable to SRM from NIST and USP