Imprimer
VWR®, Cut Resistant Gloves, NBR Coating
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
112-5679EA 51.4 EUR
112-5679 112-5680 112-5681 112-5682 112-5683 112-5684
VWR®, Cut Resistant Gloves, NBR Coating
Gants
The VWR® cut protection glove with nitrile coating provides general cut protection for the widest range of applications.
  • ANSI A2 Cut Protection (Level B)
  • HDPE/nylon/spandex blend composition
  • Nitrile (NBR) coating on palm and fingertips
  • Grey knitted glove with dark grey coating
Ideal for handling items or tools with a risk of cuts, including material handling, assembly, maintenance, and janitorial applications.
Certifications: Meets requirements for ANSI A2 Cut Resistance. EN 388:2016 (4X42B).
Certified by OKEO-TEX meeting Standard 100 certifying that no harmful substances are used in the production of these products.