Imprimer
Cartons pour b.safe Politainers
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
R299-05 R299-10 R299-20
590-0308EA 10.9 EUR
590-0308 590-0309 590-0310
Cartons pour b.safe Politainers
Boîtes Boîtes de transport
Foldable canister for the collection of liquid waste. Version as per below chart.
  • Folded to save space during delivery and storage, unfolds automatically during filling
  • Different sizes available
Certifications: UN-approval only if used with the appropriate carton box.
Informations de commande: Please order additional accessories like the carton box or the adaptor for Waste Caps GL 45 separately.