Imprimer
Waste disposal-sets, V2.0
Fournisseur: S.C.A.T.
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
307416 307315 307320 307321
SCTV307416EA 562 EUR
SCTV307416 SCTV307315 SCTV307320 SCTV307321
Waste disposal-sets, V2.0
Matériel de chromatographie Chromatographie, Pièces de rechange
These safety funnels and containers are used all over the world wherever the safe disposal of liquid waste is important. To ensure that collection points are also quickly ready for use, the most popular products have been assembled as sets. This requires only one part number for the comprehensive safety system, guaranteed compatible without time-consuming assembly of individual components.
  • Set components made of high quality and durable PE-HD material
  • Electrostatic conductive and therefore protected against ignition and explosion
  • Lid funnel with removable sieve and earthing cable
  • Available with compact 10 or large 20 L canister
  • Optional collecting tray also available as part of the set
All set components can be ordered individually under the specified article numbers.