Imprimer
Triphenylmethylium tetrakis(pentafluorophenyl)borate 97%
Marques : Apollo Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Attention
Synonyme(s): Trityltetra(pentafluorophenyl)borate, Triphenylmethylium tetrakis(perfluorophenyl)borate, Trityl tetrakis(pentafluorophenyl)borate, Triphenylcarbenium Tetrakis(pentafluorophenyl)borate, Tritylium Tetrakis(pentafluorophenyl)borate
PC56678-1G PC56678-250MG
APOSPC56678-1GEA 62.5 EUR
APOSPC56678-1G APOSPC56678-250MG
Triphenylmethylium tetrakis(pentafluorophenyl)borate 97%
Triphenylmethylium tetrakis(pentafluorophenyl)borate
Formule: C₄₃H₁₅BF₂₀
Température de stockage: Température ambiante
Numéro MDL: MFCD03426981
Numéro CAS: 136040-19-2