Imprimer

3-Fluoropyridine 98%

Marques : Apollo Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

Danger

PC4201-100G PC4201-25G PC4201-500G
APOSPC4201-100GEA 87.6 EUR
APOSPC4201-100G APOSPC4201-25G APOSPC4201-500G
3-Fluoropyridine 98%
3-Fluoropyridine

Tetrahedron, 39, 2009 (1983): with LDA lithiation is at the 2-position initially, however with longer reaction times the major product is the 4-litho derivative. Ask for other refs.

Formule: C₅H₄FN
Point d'ébullition: 106…108 °C
Densité: 1,13 g/cm³ (25 °C)
Point éclair: 13 °C
Température de stockage: Température ambiante
Numéro MDL: MFCD00006374
Numéro CAS: 372-47-4
EINECS: 206-755-4
UN: 1993
ADR: 3,II

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR