Imprimer
2,2'-(2,5-Thiènediyl)bis[5-(2-méthyl-2-propanyl)-1,3-benzoxazole] 99%
Marques : Apollo Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Attention
Synonyme(s): 2,5-Bis(5-(tert-butyl)benzo[d]oxazol-2-yl)thiophene
OR0904-100G OR0904-25G OR0904-500G
APOSOR0904-100GEA 29 EUR
APOSOR0904-100G APOSOR0904-25G APOSOR0904-500G
2,2'-(2,5-Thiènediyl)bis[5-(2-méthyl-2-propanyl)-1,3-benzoxazole] 99%
2,2'-(2,5-Thiènediyl)bis[5-(2-méthyl-2-propanyl)-1,3-benzoxazole]
Optical brightner. Highly effective in polymer substrates such as engineering plastics (polyesters, polycarbonate, polyamides and acrylics, thermoplastic polyurethane, polyvinylchloride, styrene homo- and copolymers, polyolefins), adhesives and other organic substrates.
Formule: C₂₆H₂₆N₂O₂S
Point de fusion: 196…203 °C
Température de stockage: Température ambiante
Numéro MDL: MFCD00005774
Numéro CAS: 7128-64-5