Imprimer

p-Nitrophenyl phosphate DiTris salt

Marques : Apollo Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

Attention

Synonyme(s): Phosphoric acid 4-nitrophenyl ester Bis[tris(hydroxymethyl)aminomethane] salt, 4-Nitrophenyl phosphate Bis(tris) salt, p-Nitrophenyl phosphate DiTris Salt, 4-Nitrophenyl phosphatedi(tris) salt, pNPP 2Tris

BIN4001-100G BIN4001-25G BIN4001-5G
APOSBIN4001-100GEA 284 EUR
APOSBIN4001-100G APOSBIN4001-25G APOSBIN4001-5G
p-Nitrophenyl phosphate DiTris salt
p-Nitrophenyl phosphate DiTris salt

Enzyme substrate for determination of phosphatase.

Formule: C₆H₆NO₆P
Poids moléculaire: 461.36 g/mol
Température de stockage: Réfrigérateur
Numéro MDL: MFCD00037009
Numéro CAS: 68189-42-4
EINECS: 269-198-6

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR