Imprimer
VWR® Premium 150, Premium 90 HR, Automatic Colony Counters
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
710-2073EA 14560 EUR
710-2073 710-2074
VWR® Premium 150, Premium 90 HR, Automatic Colony Counters
Colony Counters
Premium 150 and Premium 90 HR colony counters and zone measurements systems.
  • Plates can be automatically read at the press of a button
  • High definition colour images taken with a 1,4 MP scientific grade CCD camera ensure that even the smallest colony can be seen and counted, while zone measurements are fast, accurate and reproducible
  • A unique 3-channel LED lighting system configured for exceptional illumination of all sample types
  • Can be used with a new or existing stand-alone desktop or laptop computers via USB connection – no additional PCI cards are required
The Premium 150 accepts plates of up to 150 mm in diameter and detect colonies as small as 100 µm and measure zones accurately to 0,5 mm. Designed for use across a wide range of applications.
The Premium 90 HR is equipped with a high resolution 12 mm F1.4 lens, accept plates of up to 90 mm and detect colonies as small as 75 µm and measure zones accurately to 0,5 mm.
Suitable for a wide range of applications in a pharma environment.
The Premium 150 and Premium 90 HR include colony counting and inhibition zone measuring ability, spiral plate counting, Ames testing and multi-sector measurements. Other applications on demand. Results can be automatically transferred to Excel, PDF or a LIMS system. All data generated is GLP/GMP/21 CFR Part 11 compliant with a full audit trail and can be used to produce professional reports. Each system comes complete with two validation plates.
Informations de livraison: PC is not included.