Imprimer
Powder funnels, PP, SP Bel-Art Sterileware®
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F14660-1065 F14660-1080 F14660-1100 F14660-1150
231-0648EA 904 EUR
231-0648 231-0649 231-0650 231-0651
Powder funnels, PP, SP Bel-Art Sterileware®
Entonnoirs Entonnoirs à poudre
These sterile, wide stem funnels are ideal for transferring powders or granules using a sterile sampling tool into a sterile container.
  • 60° angle and large bore stem help minimise bridging of powder
  • FDA grade polypropylene; safe for food, drug and cosmetic use
  • Gamma irradiated sterile (SAL 10⁻⁶)
  • Individually bagged; ready for one time use and disposal
Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilisation.