Imprimer

Aspirin-acyl-β-D-glucuronide ≥95%

Marques : Apollo Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

Synonyme(s): Acetylsalicylic acid-acyl-β-D-glucuronide, 2-Acetoxybenzoic acid-acyl-β-D-glucuronide, ASA-acyl-β-D-glucuronide

BICL4003-1MG BICL4003-5MG BICL4003-2MG
APOSBICL4003-1MGEA 429 EUR
APOSBICL4003-1MG APOSBICL4003-5MG APOSBICL4003-2MG
Aspirin-acyl-β-D-glucuronide ≥95%
Aspirin-acyl-β-D-glucuronide
Température de stockage: Température ambiante
Numéro CAS: 24719-72-0

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR