Imprimer
Cleanroom overalls, hooded, ProShield® 60
Fournisseur: DuPont
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
P6127SWH00/S P6127SWH00/6XL P6127SWH00/2XL P6127SWH00/5XL P6127SWH00/7XL P6127SWH00/4XL P6127SWH00/3XL P6127SWH00/M P6127SWH00/L P6127SWH00/XL
DUPOP6127SWH00/SEA 409 EUR
DUPOP6127SWH00/S DUPOP6127SWH00/6XL DUPOP6127SWH00/2XL DUPOP6127SWH00/5XL DUPOP6127SWH00/7XL DUPOP6127SWH00/4XL DUPOP6127SWH00/3XL DUPOP6127SWH00/M DUPOP6127SWH00/L DUPOP6127SWH00/XL
Cleanroom overalls, hooded, ProShield® 60
Vêtements à usage unique
This overall is made of ProShield® 60 garments, it comprise a microporous film on a polypropylene nonwoven fabric and combine good particle protection with high liquid repellency. They help provide a barrier against non-hazardous dry particles down to one micron in size, are repellent to liquids. This material offers limited comfort: it is permeable to moisture vapour only and it is not air permeable.
  • Stitched internal seams
  • Nylon zipper with flap
  • Protection against radioactive contamination
  • Antistatic treatment on inside
  • Chemical protective clothing
ProShield® 60 garments are an ideal choice for applications that are less demanding in terms of barrier, durability and comfort, for example general maintenance, hospitals and other industries.
Certifications: Certified according to Regulation (EU) 2016/425; Chemical protective clothing, Category III, Type 5 and 6; EN 1073-2 (protection against radioactive contamination); Antistatic treatment (EN 1149-5).
Cdt: Packed as 50 per box, individually packed.