Imprimer
Gowns, Tyvek® IsoClean®, model IC702S 00, Non sterile
Fournisseur: DuPont
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
IC702SWH00/2XL IC702SWH00/M
DUPOIC702SWH00/2XLEA 272 EUR
DUPOIC702SWH00/2XL DUPOIC702SWH00/M
Gowns, Tyvek® IsoClean®, model IC702S 00, Non sterile
Vêtements à usage unique
These gowns are made of Tyvek® which is a fabric composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort.
  • Ultra-low linting fabric
  • Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam
  • Secure, knit cuffs for comfort and fit
  • Bound neck for lower particle shedding with ties for an adjustable fit
  • Bound ties at the waist originate at the front and tie in the back
  • Partial body chemical protective clothing
  • Barrier to infective agents
Tyvek® is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It provides an inherent barrier to particles, microorganisms and non hazardous light liquid splash. Suitable for use in GMP class C/D (ISO Class 6-9) clean rooms.
Certifications: Certified according to Regulation (EU) 2016/425; Partial body chemical protective clothing, Category III, Type PB [6-B]; EN 14126 (barrier to infective agents).
Cdt: Cardboard box 30 per box, bulk packed. 2 polyethylene liners.